Skip to Content
Merck
9-Acridinecarboxylic acid hydrate 97%

9-Acridinecarboxylic acid hydrate

9-Acridinecarboxylic acid hydrate 97%
Empirical Formula (Hill Notation):
C14H9NO2 · xH2O
CAS Number:
Molecular Weight:
223.23 (anhydrous basis)

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing