![6-Hydroxybenzothiazole-2-carbonitrile 96%](/deepweb/assets/sigmaaldrich/product/structures/246/639/d5c85699-7040-42e4-baac-aeb02fe44990/640/d5c85699-7040-42e4-baac-aeb02fe44990.png)
6-Hydroxybenzothiazole-2-carbonitrile
![6-Hydroxybenzothiazole-2-carbonitrile 96%](/deepweb/assets/sigmaaldrich/product/structures/246/639/d5c85699-7040-42e4-baac-aeb02fe44990/640/d5c85699-7040-42e4-baac-aeb02fe44990.png)
Synonym(s):
2-cyano-6-hydroxybenzothiazole
Empirical Formula (Hill Notation):
C8H4N2OS
CAS Number:
Molecular Weight:
176.20
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing