
5-(N-Methyl-N-isobutyl)amiloride

Synonym(s):
MIA
Empirical Formula (Hill Notation):
C11H18ClN7O
CAS Number:
Molecular Weight:
299.76
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing