5-Amino-2-methylphenol
Synonym(s):
3-Hydroxy-4-methylaniline, 4-Amino-2-hydroxytoluene, 5-Amino-o-cresol
Linear Formula:
H2NC6H3(CH3)OH
CAS Number:
Molecular Weight:
123.15
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing