![4-Methyl-5-(2-hydroxyethyl)thiazole British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/144/171/7b5d88a5-8e79-4898-9d67-38448cc3d182/640/7b5d88a5-8e79-4898-9d67-38448cc3d182.png)
4-Methyl-5-(2-hydroxyethyl)thiazole
![4-Methyl-5-(2-hydroxyethyl)thiazole British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/144/171/7b5d88a5-8e79-4898-9d67-38448cc3d182/640/7b5d88a5-8e79-4898-9d67-38448cc3d182.png)
Synonym(s):
4-Methyl-5-thiazoleethanol, 5-(2-Hydroxyethyl)-4-methylthiazole, Sulfurol
Empirical Formula (Hill Notation):
C6H9NOS
CAS Number:
Molecular Weight:
143.21
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing