![4-Aminopyridazine 97%](/deepweb/assets/sigmaaldrich/product/structures/980/269/b92febcb-7917-43c8-bdd6-276dfec6c6ed/640/b92febcb-7917-43c8-bdd6-276dfec6c6ed.png)
4-Aminopyridazine
![4-Aminopyridazine 97%](/deepweb/assets/sigmaaldrich/product/structures/980/269/b92febcb-7917-43c8-bdd6-276dfec6c6ed/640/b92febcb-7917-43c8-bdd6-276dfec6c6ed.png)
Empirical Formula (Hill Notation):
C4H5N3
CAS Number:
Molecular Weight:
95.10
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing