Skip to Content
Merck
4-Aminobenzylphosphonic acid ≥95%

4-Aminobenzylphosphonic acid

4-Aminobenzylphosphonic acid ≥95%
Empirical Formula (Hill Notation):
C7H10NO3P
CAS Number:
Molecular Weight:
187.13

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing