![4,4,5,5-Tetramethyl-1,3,2-dioxaborolane 97%](/deepweb/assets/sigmaaldrich/product/structures/212/620/b8a86af0-7ada-4b47-b4af-b22e74d05cab/640/b8a86af0-7ada-4b47-b4af-b22e74d05cab.png)
4,4,5,5-Tetramethyl-1,3,2-dioxaborolane
![4,4,5,5-Tetramethyl-1,3,2-dioxaborolane 97%](/deepweb/assets/sigmaaldrich/product/structures/212/620/b8a86af0-7ada-4b47-b4af-b22e74d05cab/640/b8a86af0-7ada-4b47-b4af-b22e74d05cab.png)
Synonym(s):
Pinacolborane
Empirical Formula (Hill Notation):
C6H13BO2
CAS Number:
Molecular Weight:
127.98
Products
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing