2-Propylpentanenitrile
Synonym(s):
2-Propylvaleronitrile, Dipropylacetonitrile
Linear Formula:
(CH3CH2CH2)2CHCN
CAS Number:
Molecular Weight:
125.21
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing