Skip to Content
Merck
2-Methoxy-4-methylphenol analytical standard

2-Methoxy-4-methylphenol

2-Methoxy-4-methylphenol analytical standard
Synonym(s):
2-Hydroxy-5-methylanisole, 2-Methoxy-p-cresol, 4-Hydroxy-3-methoxytoluene, 4-Methylguaiacol, Creosol
Linear Formula:
CH3OC6H3(CH3)OH
CAS Number:
Molecular Weight:
138.16
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-4 of 4