2-Isopropylmalic acid
Synonym(s):
2-Hydroxy-2-isopropylsuccinic acid
Linear Formula:
HO2CCH2C[CH(CH3)2](OH)CO2H
CAS Number:
Molecular Weight:
176.17
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing