![2-Arachidonyl glycerol ~10 mg/mL, ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/232/592/ab43361e-b020-4313-a9c6-dc379d4aa71e/640/ab43361e-b020-4313-a9c6-dc379d4aa71e.png)
2-Arachidonyl glycerol
![2-Arachidonyl glycerol ~10 mg/mL, ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/232/592/ab43361e-b020-4313-a9c6-dc379d4aa71e/640/ab43361e-b020-4313-a9c6-dc379d4aa71e.png)
Synonym(s):
(all-Z)-5,8,11,14-Eicosatetraenoic acid 2-hydroxy-1-(hydroxymethyl)ethyl ester, 2-AG, 2-Arachidonoylglycerol, 2-Monoarachidonoylglycerol
Empirical Formula (Hill Notation):
C23H38O4
CAS Number:
Molecular Weight:
378.55
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing