Skip to Content
Merck
2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol 97%

2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol

2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol 97%
Synonym(s):
Bromo-PADAP
Empirical Formula (Hill Notation):
C15H17BrN4O
CAS Number:
Molecular Weight:
349.23
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing