Skip to Content
Merck
2,3-Hexanedione analytical standard

2,3-Hexanedione

2,3-Hexanedione analytical standard
Synonym(s):
Acetyl butyryl, 2,3-Hexanedione
Linear Formula:
CH3CH2CH2COCOCH3
CAS Number:
Molecular Weight:
114.14
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5