1H,1H,2H,2H-Perfluoro-1-octanol
Synonym(s):
3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanol, 1H,1H,2H,2H-Perfluoro-1-octanol
Linear Formula:
CF3(CF2)5CH2CH2OH
CAS Number:
Molecular Weight:
364.10
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Technique
Showing 1-2 of 2