Skip to Content
Merck
17β-Hydroxy-17-methylandrosta-1,4-dien-3-one analytical standard

17β-Hydroxy-17-methylandrosta-1,4-dien-3-one

17β-Hydroxy-17-methylandrosta-1,4-dien-3-one analytical standard
Synonym(s):
1-Dehydro-17α-methyltestosterone, Dianabol, Methandienone, Methandrostenolone
Empirical Formula (Hill Notation):
C20H28O2
CAS Number:
Molecular Weight:
300.44
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing