Skip to Content
Merck
1-[(2RS)-2-(Benzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazolium nitrate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

1-[(2RS)-2-(Benzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazolium nitrate

1-[(2RS)-2-(Benzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazolium nitrate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Miconazole Nitrate impurity H (PhEur)
Empirical Formula (Hill Notation):
C18H16Cl2N2O · HNO3
CAS Number:
Molecular Weight:
410.25

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade