Skip to Content
Merck
1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid ≥96.0% (HPLC)

1,2-Bis(2-Aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid

1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid ≥96.0% (HPLC)
Synonym(s):
2,2′-(Ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid, BAPTA, Ethylenedioxybis(o-phenylenenitrilo)tetraacetic acid, Ethylenedioxybis(o-phenylenenitrilo)tetraacetic acid
Empirical Formula (Hill Notation):
C22H24N2O10
CAS Number:
Molecular Weight:
476.43

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group