Skip to Content
Merck
1,2:5,6-Di-O-cyclohexylidene-D-mannitol 98%

1,2:5,6-Di-O-cyclohexylidene-D-mannitol

1,2:5,6-Di-O-cyclohexylidene-D-mannitol 98%
Empirical Formula (Hill Notation):
C18H30O6
CAS Number:
Molecular Weight:
342.43

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing