Skip to Content
Merck
1,1,1,2,2,3,3,7,7,8,8,9,9,9-Tetradecafluoro-4,6-nonanedione 97%

1,1,1,2,2,3,3,7,7,8,8,9,9,9-Tetradecafluoro-4,6-nonanedione

1,1,1,2,2,3,3,7,7,8,8,9,9,9-Tetradecafluoro-4,6-nonanedione 97%
Synonym(s):
5H,5H-Perdecafluoro-4,6-nonandione
Linear Formula:
(CF3CF2CF2CO)2CH2
CAS Number:
Molecular Weight:
408.09

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing