Product Name
Hexakis[(4-methylphenyl)thio]benzene 98%
Product Number
495948
Product Brand
aldrich
CAS Number
55709-41-6
Molecular Weight
811.24
Test | Specification |
---|---|
APPEARANCE | YELLOW TO GREEN POWDER, CRYSTALS, |
CRYSTALLINE POWDER AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
ELEMENTAL ANALYSIS | CARBON 69.3%-72.8% |
SULFUR 23.1%-24.3% | |
HIGH PRESSURE LIQUID | 97.5% (MINIMUM) |
APPROVED JULY 11, 2007 RJM |