Spectroquant® Prove Connect to LIMS
![Spectroquant® Photometers a modern, compact, green and black kitchen appliance with a touchscreen interface on the lid. The device has a prominent yellow “M” logo on the side and is accompanied by two additional lids in yellow and black colors, each featuring the same logo](/deepweb/assets/sigmaaldrich/marketing/global/images/categories/chemical-synthesis/spectroquant-photometers/spectroquant-photometers.jpg)
Integrate your Prove Photometer into your Lab System
Automate and streamline quality control data or experimental data transfer to your electronic lab system with Spectroquant® Prove Connect to LIMS software.
- Allows an easy link of the data generated by you Prove spectrophotometer to your LIMS (Laboratory Information Management System) or ELN (Electronic Lab Notebook) by simply connecting via LAN.
- Offers flexible integration into other systems such as ELNs (Electronic Lab Notebook).
Order your Spectroquant® Prove Connect to LIMS unlimited license or get a free trial license
Our Spectroquant® Prove spectrophotometer series offers easy analyses of drinking, wastewater, and process water - with over 200 pre-programmed Spectroquant® test kits for your specific application.
The digital solutions from the Supelco® portfolio can simplify your daily analytical workflow and maintain the fidelity of your data while saving you time for your important tasks.
Request Trial License
Please login to your account to obtain a license key.
Login
Get Started
Instruction for download and installation:
- Get a license key**.
- Download Spectroquant® Prove Connect to LIMS software & manual for free.
Software: Spectroquant® Prove Connect to LIMS Software Version 2.0
Manual: Spectroquant® Prove Connect to LIMS
- Install the software as described below:
- Install the software by unzipping the file and opening the *.exe.
- When starting the software for the first time, enter your license key when requested.
- Please follow the instructions in the manual to complete the installation using your local IT infrastructure.
**Please have the serial number(s) of your Spectroquant® Prove spectrophotometer(s) at hand.
*** Trial license valid for xxx
Compatibility
The software supports CSV and XML data formats and is compatible with the following photometers as well as their predecessor Spectroquant® Prove 600/300/100:
Computer requirements*:
- Microsoft Windows 7 SP1, Windows 8 or Windows 10
- 1 GB of free disk space
- 4 GB of RAM
- Network Connection (LAN)
*Other configurations may work but are not supported by Merck