Skip to Content
Merck
All Photos(16)

Documents

ZRB2234

Sigma-Aldrich

Anti-HES-1 Antibody, clone 2J12 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Class B basic helix-loop-helix protein 39, Hairy and enhancer of split 1, Hairy homolog, Hairy-like protein, Transcription factor HES-1, bHLHb39, hHL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2J12, recombinant monoclonal

description

2J12 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 29.54 kDa
observed mol wt ~34 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

porcine, feline, bovine, monkey, canine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... HES1(3280)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2J12 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Transcription factor HES-1. It targets an epitope within 15 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal region of human Transcription factor HES-1.

Application

Quality Control Testing

Evaluated by Western Blotting in HEK293 cell nuclear extract.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected HES-1 in HEK293 cell nuclear extract.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected HES-1 in SK-BR3 cell nuclear extract.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected HES-1 in Human thyroid tissue sections.

Flow Cytometry Analysis: 0.1 µg from a representative lot detected HES-1 in one million SK-BR-3 cells.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected HES-1 in MCF-7 and SK-BR-3 cells.

Affinity Binding Assay: A representative lot of this antibody bound HES-1 peptide with a KD of 7.0 x 10-9 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Transcription factor HES-1 (UniProt: Q14469; also known as Class B basic helix-loop-helix protein 39, bHLHb39, Hairy and enhancer of split 1, Hairy homolog, Hairy-like protein, hHL) is encoded by the HES1 (also known as BHLHB39, HL, HRY) gene (Gene ID: 3280) in human. HES-1 acts as a transcriptional repressor of genes that require basic helix-loop-helix (bHLH) protein for their transcription. It plays a role in several physiological processes, including cell differentiation, cell cycle arrest, and apoptosis. It is expressed in high levels in undifferentiated neural precursor cells and its levels decline with neural differentiation. It also acts as a negative regulator of myogenesis by inhibiting the function of Myogenic Differentiation 1 (MYOD1) and transcription regulatory protein ASH1. HES-1 also serves as a relevant mediator of the NOTCH signaling pathway in cancer where NOTCH pathway, through HES-1, is reported to contribute to the stabilization of the epithelial phenotype of bladder cancer cells. Low levels of HES-1 correlate with more aggressive bladder cancer and associate with mesenchymal features and invasive properties. However, HES1 levels are upregulated in poorly differentiated cancers compared with well-differentiated tumor samples and most adenocarcinomas display higher levels of HES1 mRNA. Its expression is correlated with the expression of stem cell markers in colon cancers, and it upregulates the expression of stemness-related genes in colon cancer cells. HES-1 contains a bHLH domain (aa 34-91), Orange domain (aa 110-143), and a C-terminal WRPW (Trp-Arg-Pro-Trp) motif (aa 275-278).  The bHLH, as well as cooperation between the central Orange domain and the C-terminal WRPW motif, are reported to be essential for its transcriptional repressor activity. Within the bHLH region, HES-1 has a proline residue that exhibits unique binding ability to the N box (CACNAG), differing from other bHLH factors that have higher affinity to E box (CANNTG).  This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Maraver, A et al., (2015). J. Clin. Invest. 125(2); 824-830; Liu, Z-H., et al. (2015). Cancer Biol. Ther. 16(3); 353-359; Gao, F., et al. (2014). Sci. Rep. 4; Article 3963). 

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service