Skip to Content
Merck
All Photos(1)

Documents

I7534

Sigma-Aldrich

Monoclonal Anti-Interleukin-1α antibody produced in mouse

clone 4414, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-IL-1α

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4414, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

capture ELISA: 2-8 μg/mL
immunohistochemistry: 8-25 μg/mL
neutralization: suitable
western blot: 1-2 μg/mL

isotype

IgG2a

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL1A(3552)

Looking for similar products? Visit Product Comparison Guide

Immunogen

purified, E. coli-derived recombinant human interleukin-1α (aa 113-271).

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing carbohydrates.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service