Skip to Content
Merck
All Photos(1)

Key Documents

73B

91060554, Reactivity: Anti-CD45RA, ovine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

Sp2/0

Reactivity

Anti-CD45RA, ovine

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Sp2/0 x Mouse BALB/c spleen

Cell Line Description

The antibody is directed against the p220 component of the sheep CD45 molecule (CD45RA), expressed on all B cells and a sub-population of naive T cells.

Immunogen

Sheep lymphocytes

Application

Useful for distinguishing naive and memory T cells, FACS

Culture Medium

EMEM(EBSS) + 2mM glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service