Skip to Content
Merck
All Photos(1)

Documents

A1685

Sigma-Aldrich

2-Methyl-2-butanol

≥99%

Synonym(s):

tert-Amyl alcohol, tert-Pentyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2C(CH3)2OH
CAS Number:
Molecular Weight:
88.15
Beilstein:
1361351
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor density

3 (vs air)

Assay

≥99%

autoignition temp.

819 °F

expl. lim.

9 %

refractive index

n20/D 1.405 (lit.)

bp

102 °C (lit.)

mp

−12 °C (lit.)

density

0.805 g/mL at 25 °C (lit.)

SMILES string

CCC(C)(C)O

InChI

1S/C5H12O/c1-4-5(2,3)6/h6H,4H2,1-3H3

InChI key

MSXVEPNJUHWQHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

68.9 °F - closed cup

Flash Point(C)

20.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service