Skip to Content
Merck
All Photos(1)

Key Documents

81640

Sigma-Aldrich

5-Pregnen-3β-ol-20-one acetate

purum

Synonym(s):

3β-Acetoxy-5-pregnen-20-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H34O3
CAS Number:
Molecular Weight:
358.51
Beilstein:
2062850
EC Number:
MDL number:
UNSPSC Code:
12352000

grade

purum

optical activity

[α]20/D +19±2°, c = 1% in ethanol

mp

148-150 °C

SMILES string

CC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CC[C@@H]4C(C)=O)[C@@H]3CC=C2C1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service