Skip to Content
Merck
All Photos(2)

Key Documents

04535

Supelco

4(5)-Methylimidazole

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2
CAS Number:
Molecular Weight:
82.10
Beilstein:
1453
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

bp

263 °C (lit.)

mp

44-47 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
forensics and toxicology
personal care
veterinary

format

neat

storage temp.

2-8°C

SMILES string

Cc1c[nH]cn1

InChI

1S/C4H6N2/c1-4-2-5-3-6-4/h2-3H,1H3,(H,5,6)

InChI key

XLSZMDLNRCVEIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matt Hengel et al.
Journal of agricultural and food chemistry, 61(4), 780-789 (2013-01-09)
Since the National Toxicology Program (NTP) identified 4(5)-methylimidazole [4(5)-MI] as a cancer causing chemical in 2007 and the State of California added it to the Proposition 65 list of compounds as a carcinogen on January 7, 2011, many researchers and
Julie Kim et al.
Talanta, 129, 106-112 (2014-08-17)
Using the role of p-iodophenol in enzyme assay, enhanced 1,1'-oxalyldiimidazole chemiluminescent enzyme immunoassays (ODI-CLEIAs) were developed to consecutively quantify trace levels of triple tumor markers, such as alpha fetoprotein (AFP), carcinoembryonic antigen (CEA), and prostate specific antigen (PSA) in a
Sabine Gerbal-Chaloin et al.
Molecular pharmacology, 86(6), 624-634 (2014-09-18)
The wingless-type MMTV integration site family (WNT)/β-catenin/adenomatous polyposis coli (CTNNB1/APC) pathway has been identified as a regulator of drug-metabolizing enzymes in the rodent liver. Conversely, little is known about the role of this pathway in drug metabolism regulation in human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service