Skip to Content
Merck
All Photos(1)

Key Documents

11585738910

Roche

DIG-labeled Control DNA

greener alternative

solution, pkg of 50 μL (5 μg/ml DIG-labeled plasmid DNA)

Synonym(s):

control DNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105330

form

solution

packaging

pkg of 50 μL (5 μg/ml DIG-labeled plasmid DNA)

manufacturer/tradename

Roche

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

−20°C

General description

Determination of the yield of digoxigenin (DIG)-labeled DNA is most important for optimal and reproducible hybridization results. Too high of a probe concentration in the hybridization step causes background, while too low of a concentration leads to weak signals.
DIG-labeled control has a standardized concentration and can be used as a standard for comparison to experimentally generated DIG-labeled probes.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemical.  The DIG System was established as a sensitive and cost-effective alternative to radioactivity for the labeling and detection of nucleic acids. There are many available publications that prove the versatility of the DIG System, so use of radio-labeling is no longer the only option for labeling of DNA for hybridization.

Application

Estimation of the yield of DIG-labeled nucleic acids or as a control-hybridization probe in Southern blots.

Quality

0.1pg of the DIG-labeled Control DNA can be visualized in a spot test with direct color detection.

Physical form

Solution, digoxigenin-labeled pBR328 DNA, 5 μg/ml

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Digoxigenin (DIG) labeling methods and kits for DNA and RNA DIG probes, random primed DNA labeling, nick translation labeling, 5’ and 3’ oligonucleotide end-labeling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service