Skip to Content
Merck
All Photos(1)

Documents

2610-OP

Millipore

N,N′-Methylene Bis Acrylamide

OmniPur® Grade, >= 99.0%, Suitable for nucleic acid and protein separation, sequencing.

Synonym(s):

OmniPur® N,N′-Methylene Bis Acrylamide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41105319

Quality Level

form

crystalline powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C7H10N2O2/c1-3-6(10)8-5-9-7(11)4-2/h3-4H,1-2,5H2,(H,8,10)(H,9,11)

InChI key

ZIUHHBKFKCYYJD-UHFFFAOYSA-N

General description

Suitable for nucleic acid and protein separation, sequencing - standard resolution. Cross-linker for PAGE gels used in various ratios to achieve different gel strengths. Molecular biology grade.

Specifications:
A (290 nm, 1%, water): ≤0.2AU
Acrylic acid: ≤0.001%
Assay: ≥99.0%
Conductivity: ≤10 µmho
DNase: None detected
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 1B - Muta. 1B - Repr. 2 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service