ENAH3045DED3
Enamine
N-[3-(morpholin-4-yl)propyl]cyclopentanamine
About This Item
Iupac Name
N-[3-(4-morpholinyl)propyl]cyclopentanamine
Inchi Code
1S/C12H24N2O/c1-2-5-12(4-1)13-6-3-7-14-8-10-15-11-9-14/h12-13H,1-11H2
InChI key
LNLJOSKUGCECFZ-UHFFFAOYSA-N
Country of Origin
Product Link
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service