Skip to Content
Merck
All Photos(1)

Key Documents

COMH6E40A362

Combi-Blocks, Inc.

2,5-Diamino-4,6-dihydroxypyrimidine hydrochloride monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€156.92
5 G
€364.59
25 G
€987.62

Vendor SKU: OR-7211-1G

€156.92


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
€156.92
5 G
€364.59
25 G
€987.62

About This Item

CAS Number:
Molecular Weight:
196.59
MDL number:

Vendor SKU: OR-7211-1G

€156.92


Ships in 14 days - from Aldrich Partner.

Iupac Name

2,5-diamino-4,6-pyrimidinediol hydrochloride hydrate

Inchi Code

1S/C4H6N4O2.ClH.H2O/c5-1-2(9)7-4(6)8-3(1)10;;/h5H2,(H4,6,7,8,9,10);1H;1H2

InChI key

WNMOLDIYHAYRNV-UHFFFAOYSA-N

storage temp.

Freezer

Purity

98%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?OR-7211

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?OR-7211


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service