CIAH987EDBF2
ChemScene LLC (Preferred Partner)
3-Amino-5-methylbenzonitrile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
3-amino-5-methylbenzonitrile
Inchi Code
1S/C8H8N2/c1-6-2-7(5-9)4-8(10)3-6/h2-4H,10H2,1H3
InChI key
UNPIGVYJSNRGBR-UHFFFAOYSA-N
storage temp.
4C, protect from light
Purity
97%
Country of Origin
CN
Physical Form
Solid
Product Link
https://www.chemscene.com/186551-97-3.html
Signal Word
Danger
Hazard Statements
Precautionary statements
MSDS
https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0198299.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service