Skip to Content
Merck
All Photos(1)

Key Documents

ATC386058620

Atlantic Research Chemicals Ltd

Ethyl 5-Bromonicotinate 1-oxide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
246.06
MDL number:

Iupac Name

ethyl 5-bromonicotinate 1-oxide

Inchi Code

1S/C8H8BrNO3/c1-2-13-8(11)6-3-7(9)5-10(12)4-6/h3-5H,2H2,1H3

InChI key

YHVZWNYKNZDWRU-UHFFFAOYSA-N

Purity

95

Country of Origin

GB

Product Link

https://atlantic-chemicals.com/product/?code=ES002108

MSDS

https://atlantic-chemicals.com/product/?code=ES002108


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service