Skip to Content
Merck
All Photos(1)

Key Documents

ATE429355117

AstaTech, Inc.

2-(OXETAN-3-YL)ACETALDEHYDE

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€428.76
250 MG
€921.32
1 G
€2,558.76

Vendor SKU: CS2316-50MG

€428.76


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
€428.76
250 MG
€921.32
1 G
€2,558.76

About This Item

CAS Number:
Molecular Weight:
100.12
MDL number:

Vendor SKU: CS2316-50MG

€428.76


Ships in 14 days - from Aldrich Partner.

Iupac Name

2-(oxetan-3-yl)acetaldehyde

Inchi Code

1S/C5H8O2/c6-2-1-5-3-7-4-5/h2,5H,1,3-4H2

InChI key

AIFBKSGZBRGQJE-UHFFFAOYSA-N

storage temp.

-20 C

Purity

95%

Shipping Temperature

Normal

Physical Form

Colorless to Yellow Sticky Oil to Semi-Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=45675

Signal Word

Warning

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=CS2316


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service