Skip to Content
Merck
All Photos(1)

Documents

688991

Sigma-Aldrich

2-Methylbutyronitrile

Lonza quality, ≥80.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9N
CAS Number:
Molecular Weight:
83.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

Lonza quality

Assay

≥80.0% (GC)

form

liquid

manufacturer/tradename

Lonza Ltd

bp

125-126 °C

SMILES string

CCC(C)C#N

InChI

1S/C5H9N/c1-3-5(2)4-6/h5H,3H2,1-2H3

InChI key

RCEJCSULJQNRQQ-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

77.0 °F

Flash Point(C)

25 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Cutler et al.
Archives of biochemistry and biophysics, 238(1), 272-279 (1985-04-01)
Microsomal preparations from flax seedlings have recently been shown to convert L-valine to acetone cyanohydrin, the precursor of the cyanogenic glucoside linamarin [A. J. Cutler and E. E. Conn (1981) Arch. Biochem. Biophys. 212, 468-474]. Further details of this four-step
Kartheek Katta et al.
Beilstein journal of nanotechnology, 10, 522-530 (2019-03-16)
We report the synthesis of high-performance organic-inorganic hybrid fluorescent nanocapsules comprising a polymer shell armored with an inorganic layer and a liquid core containing a fluorophore. The polymeric capsules are synthesized by free radical miniemulsion polymerization and contain covalently bound
Maria Kokkinopoulou et al.
Nanoscale, 9(25), 8858-8870 (2017-06-21)
The use of nanocarriers in biology and medicine is complicated by the current need to understand how nanoparticles interact in complex biological surroundings. When nanocarriers come into contact with serum, proteins immediately adsorb onto their surface, forming a protein corona
Sandra Ritz et al.
Biomacromolecules, 16(4), 1311-1321 (2015-03-21)
Understanding nanoparticle-protein interactions is a crucial issue in the development of targeted nanomaterial delivery. Besides unraveling the composition of the nanoparticle's protein coronas, distinct proteins thereof could control nanoparticle uptake into specific cell types. Here we differentially analyzed the protein
Phillip Trefz et al.
PloS one, 8(10), e76868-e76868 (2013-10-12)
Control of paratuberculosis in livestock is hampered by the low sensitivity of established direct and indirect diagnostic methods. Like other bacteria, Mycobacterium avium subsp. paratuberculosis (MAP) emits volatile organic compounds (VOCs). Differences of VOC patterns in breath and feces of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service