Skip to Content
Merck
All Photos(1)

Documents

463663

Sigma-Aldrich

Barium perchlorate

99.999%

Synonym(s):

Barium diperchlorate, Perchloric acid barium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba(ClO4)2
CAS Number:
Molecular Weight:
336.23
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99.999%

reaction suitability

reagent type: oxidant

mp

505 °C (lit.)

density

3.2 g/mL at 25 °C (lit.)

SMILES string

[Ba++].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/Ba.2ClHO4/c;2*2-1(3,4)5/h;2*(H,2,3,4,5)/q+2;;/p-2

InChI key

OOULUYZFLXDWDQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Investigations of:
  • Effects on cationic Gemini surfactant monolayers

Catalyst for:
  • Barium cation-templated synthesis and cation induced molecular pirouetting
  • Olefin dioxygenation

Used for:
  • Cation-induced molecular motion of spring-like [2]catenanes
  • Complexation of tweezer molecule with barium ion
  • Preparation of Schiff base macrocyclic barium complexes and studies of antibacterial activity

Reactant for oxidation of bis(amine)

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

69.8 °F - closed cup

Flash Point(C)

21 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C Gallucci et al.
Acta crystallographica. Section C, Crystal structure communications, 44 ( Pt 11), 1873-1876 (1988-11-15)
Ba(ClO4)2.3H2O, Mr = 390.28, hexagonal, P63/m, a = 7.277 (2), c = 9.656 (1) A, V = 442.9 (1) A3, Z = 2, F(000) = 368, D chi = 2.93 g cm3, lambda(Mo K alpha) = 0.71069 A, T =

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service