Skip to Content
Merck
All Photos(1)

Key Documents

371971

Sigma-Aldrich

2,2,3,4,4,4-Hexafluorobutyl methacrylate

98%

Synonym(s):

HFBMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)COOCH2CF2CFHCF3
CAS Number:
Molecular Weight:
250.14
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

0.25 psi ( 20 °C)

Assay

98%

form

liquid

contains

hydroquinone as stabilizer

refractive index

n20/D 1.361 (lit.)

bp

158 °C (lit.)

density

1.348 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCC(F)(F)C(F)C(F)(F)F

InChI

1S/C8H8F6O2/c1-4(2)5(15)16-3-7(10,11)6(9)8(12,13)14/h6H,1,3H2,2H3

InChI key

DFVPUWGVOPDJTC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

G Kristin Jonsson et al.
Colloid and polymer science, 295(10), 1983-1991 (2017-10-11)
Fluorinated spheres with grafted poly(ethylene glycol) (PEG) have been synthesized using a semi-batch emulsion polymerization in which the initiator is fed slowly to the reaction. In this way, PEG-grafted colloidal spheres can be fabricated with varying PEG chain length, different

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service