Skip to Content
Merck
All Photos(1)

Documents

300411

Sigma-Aldrich

Triton X-114, reduced

reduced

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H60O9
Molecular Weight:
564.79
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

liquid

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI key

HNLXNOZHXNSSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sho Yui et al.
The Japanese journal of veterinary research, 62(4), 151-162 (2015-01-20)
The mechanism by which olfactory ensheathing cells (OECs) exert their potential to promote functional recovery after transplantation into spinal cord injury (SCI) tissue is not fully understood, but the relevance of matrix metalloproteinases (MMPs) has been suggested. We evaluated the
Flemming Jessen et al.
Analytical biochemistry, 485, 11-17 (2015-06-17)
A simple and reproducible procedure for enrichment of a plasma protein subfraction suitable for two-dimensional polyacrylamide gel electrophoresis (2DE) was developed, using a Triton X-114-based cloud point extraction (CPE). Appropriate conditions for such a CPE procedure were found by SDS-PAGE
Adam J Blanch et al.
Small (Weinheim an der Bergstrasse, Germany), 11(35), 4550-4559 (2015-06-13)
Branched gold nanoparticles with sharp tips are considered excellent candidates for sensing and field enhancement applications. Here, a rapid and simple synthesis strategy is presented that generates highly branched gold nanoparticles with hollow cores and a ca.100% yield through a
Celia Fernández-Blanco et al.
Toxicology letters, 235(2), 61-66 (2015-04-02)
Oxidative stress occurs as a consequence of an imbalance between the prooxidant/antioxidant systems, causing an increase of intracellular generation of reactive oxygen species. Alternariol (AOH), a mycotoxin produced by Alternaria sp. can alter the action of glutathione (GSH) and the
Mohammad Yaqoob et al.
Luminescence : the journal of biological and chemical luminescence, 30(7), 950-961 (2015-02-03)
A flow-injection (FI) method is reported for the determination of Mn(II), maneb and mancozeb fungicides based on the catalytic effect of Mn(II) on the oxidation of lucigenin and dissolved oxygen in a basic solution. The Tween-20 surfactant has been reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service