Skip to Content
Merck
All Photos(1)

Key Documents

232025

Sigma-Aldrich

4-(Trifluoromethyl)benzyl alcohol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4CH2OH
CAS Number:
Molecular Weight:
176.14
Beilstein:
1450174
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.459 (lit.)

bp

78-80 °C/4 mmHg (lit.)

density

1.286 g/mL at 25 °C (lit.)

SMILES string

OCc1ccc(cc1)C(F)(F)F

InChI

1S/C8H7F3O/c9-8(10,11)7-3-1-6(5-12)2-4-7/h1-4,12H,5H2

InChI key

MOOUWXDQAUXZRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Photocatalytic oxidation of 4-(trifluoromethyl)benzyl alcohol to corresponding aldehydes on TiO2 photocatalyst under O2 atmosphere has been reported.

Application

4-(Trifluoromethyl)benzyl alcohol was used:
  • in the synthesis of highly electron-poor fluorine-containing alkenes
  • as a reagent in kinetic studies of phosphonoformate prodrugs and aquachromium (IV)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Chem. Soc. Perkin Trans. II, 1145-1145 (1992)
One-pot synthesis of fluorine-containing alkenes from in situ-generated stabilized phosphorus ylides.
Pakravan P, et al.
Phosphorus, Sulfur, and Silicon and the Related Elements, 182(3), 545-549 (2007)
Journal of the American Chemical Society, 114, 4205-4205 (1992)
Selective photocatalytic oxidation of benzyl alcohol and its derivatives into corresponding aldehydes by molecular oxygen on titanium dioxide under visible light irradiation.
Higashimoto S, et al.
J. Catal., 266(2), 279-285 (2009)
E Baciocchi et al.
European journal of biochemistry, 268(3), 665-672 (2001-02-13)
The H(2)O(2)-promoted oxidations of (R)-[alpha-(2)H(1)]-and (S)-[alpha-(2)H(1)]-arylalkanols catalysed by chloroperoxidase (CPO) from Caldariomyces fumago have been investigated. It has been found that with (R)-[alpha-(2)H(1)]-alcohols, the oxidation involves almost exclusively the cleavage of the C-H bond, whereas in the case of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service