Skip to Content
Merck
All Photos(2)

Key Documents

220434

Sigma-Aldrich

3-Hydroxybenzophenone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4COC6H5
CAS Number:
Molecular Weight:
198.22
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

mp

113-115 °C (lit.)

SMILES string

Oc1cccc(c1)C(=O)c2ccccc2

InChI

1S/C13H10O2/c14-12-8-4-7-11(9-12)13(15)10-5-2-1-3-6-10/h1-9,14H

InChI key

SHULEACXTONYPS-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Marina G Pintado-Herrera et al.
Journal of chromatography. A, 1429, 107-118 (2016-01-10)
The main goal of this work was to develop, optimize and validate a multi-residue method for the simultaneous determination of 97 contaminants, including fragrances, UV filters, repellents, endocrine disruptors, biocides, polycyclic aromatic hydrocarbons (PAHs), polychlorinated biphenyls (PCBs), organophosphorus flame retardants
Marina G Pintado-Herrera et al.
Analytica chimica acta, 851, 1-13 (2014-12-03)
This work presents the development, optimization and validation of a multi-residue method for the simultaneous determination of 102 contaminants, including fragrances, UV filters, repellents, endocrine disruptors, biocides, polycyclic aromatic hydrocarbons (PAHs), polychlorinated biphenyls (PCBs), and several types of pesticides in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service