Skip to Content
Merck
All Photos(1)

Key Documents

AC7300012910

Advanced Chemical Intermediates Ltd.

Ethyl 2-methyl-4-(trifluoromethyl)thiazole-5-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
239.22
MDL number:

Iupac Name

ethyl 2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate

Inchi Code

1S/C8H8F3NO2S/c1-3-14-7(13)5-6(8(9,10)11)12-4(2)15-5/h3H2,1-2H3

InChI key

FKNJDCSKTSRSSW-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Room temperature

Physical Form

Yellow oil

Product Link

https://www.acints.com/products/ACI-NI-00055

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.acints.com/products/ACI-NI-00055


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service