Skip to Content
Merck
All Photos(1)

Key Documents

ADVH7F37B92B

AChemBlock

4-bromo-2,3-dihydrobenzofuran-3-amine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€311.07
250 MG
€482.54
1 G
€1,125.53

Vendor SKU: T94414-100MG

€311.07


Ships in 21 days - from Aldrich Partner.


Select a Size

Change View
100 MG
€311.07
250 MG
€482.54
1 G
€1,125.53

About This Item

CAS Number:
Molecular Weight:
214.06
MDL number:

Vendor SKU: T94414-100MG

€311.07


Ships in 21 days - from Aldrich Partner.

Iupac Name

4-bromo-2,3-dihydrobenzofuran-3-amine

Inchi Code

1S/C8H8BrNO/c9-5-2-1-3-7-8(5)6(10)4-11-7/h1-3,6H,4,10H2

InChI key

FDWXLXRSLLDBEX-UHFFFAOYSA-N

storage temp.

Store at 0-8 C

Purity

97%

Country of Origin

CN

Physical Form

yellow oil

Product Link

https://www.achemblock.com/t94414-4-bromo-2-3-dihydrobenzofuran-3-amine.html

Synonym

4-bromo-2,3-dihydrobenzofuran-3-amine

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.achemblock.com/t94414-4-bromo-2-3-dihydrobenzofuran-3-amine.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service