Skip to Content
Merck
All Photos(1)

Key Documents

AC5372908318

Accela ChemBio Inc.

1-(2-Fluoro-4-nitrophenyl)pyrrolidine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€145.73
5 G
€360.06

Vendor SKU: SY002977-1G

€145.73


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
€145.73
5 G
€360.06

About This Item

CAS Number:
Molecular Weight:
210.21
MDL number:

Vendor SKU: SY002977-1G

€145.73


Ships in 14 days - from Aldrich Partner.

Iupac Name

1-(2-fluoro-4-nitrophenyl)pyrrolidine

Inchi Code

1S/C10H11FN2O2/c11-9-7-8(13(14)15)3-4-10(9)12-5-1-2-6-12/h3-4,7H,1-2,5-6H2

InChI key

SMKXLBISYVJESG-UHFFFAOYSA-N

storage temp.

RT

Purity

97%

Country of Origin

CN

Shipping Temperature

RT

Physical Form

Yellow to orange powder

Product Link

http://www.accelachem.com/cn/search.html?keyword=SY002977&attname=GoodsCode

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

http://pdf.accelachem.com/cn/msds/SY002977.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service