Merck
  • CRISPR-Cas9 screen reveals a MYCN-amplified neuroblastoma dependency on EZH2.
CRISPR-Cas9 screen reveals a MYCN-amplified neuroblastoma dependency on EZH2.
The Journal of clinical investigation (2017-12-05)
Liying Chen, Gabriela Alexe, Neekesh V Dharia, Linda Ross, Amanda Balboni Iniguez, Amy Saur Conway, Emily Jue Wang, Veronica Veschi, Norris Lam, Jun Qi, W Clay Gustafson, Nicole Nasholm, Francisca Vazquez, Barbara A Weir, Glenn S Cowley, Levi D Ali, Sasha Pantel, Guozhi Jiang, William F Harrington, Yenarae Lee, Amy Goodale, Rakela Lubonja, John M Krill-Burger, Robin M Meyers, Aviad Tsherniak, David E Root, James E Bradner, Todd R Golub, Charles Wm Roberts, William C Hahn, William A Weiss, Carol J Thiele, Kimberly Stegmaier
ABSTRACT
Pharmacologically difficult targets, such as MYC transcription factors, represent a major challenge in cancer therapy. For the childhood cancer neuroblastoma, amplification of the oncogene MYCN is associated with high-risk disease and poor prognosis. Here, we deployed genome-scale CRISPR-Cas9 screening of MYCN-amplified neuroblastoma and found a preferential dependency on genes encoding the polycomb repressive complex 2 (PRC2) components EZH2, EED, and SUZ12. Genetic and pharmacological suppression of EZH2 inhibited neuroblastoma growth in vitro and in vivo. Moreover, compared with neuroblastomas without MYCN amplification, MYCN-amplified neuroblastomas expressed higher levels of EZH2. ChIP analysis showed that MYCN binds at the EZH2 promoter, thereby directly driving expression. Transcriptomic and epigenetic analysis, as well as genetic rescue experiments, revealed that EZH2 represses neuronal differentiation in neuroblastoma in a PRC2-dependent manner. Moreover, MYCN-amplified and high-risk primary tumors from patients with neuroblastoma exhibited strong repression of EZH2-regulated genes. Additionally, overexpression of IGFBP3, a direct EZH2 target, suppressed neuroblastoma growth in vitro and in vivo. We further observed strong synergy between histone deacetylase inhibitors and EZH2 inhibitors. Together, these observations demonstrate that MYCN upregulates EZH2, leading to inactivation of a tumor suppressor program in neuroblastoma, and support testing EZH2 inhibitors in patients with MYCN-amplified neuroblastoma.
Sigma-Aldrich
PF-06463922 acetate, ≥98% (HPLC)
Sigma-Aldrich
Anti-Histone H3 Antibody, 0.5 mg/mL, UpstateĀ®
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, UpstateĀ®, from rabbit