Merck
Cell dynamics underlying oriented growth of the
Development (Cambridge, England) (2017-10-19)
Natalie A Dye, Marko Popović, Stephanie Spannl, Raphaël Etournay, Dagmar Kainmüller, Suhrid Ghosh, Eugene W Myers, Frank Jülicher, Suzanne Eaton
ABSTRACT
Quantitative analysis of the dynamic cellular mechanisms shaping the
Sigma-Aldrich
Insulin from bovine pancreas, ≥25 USP units/mg (HPLC), powder
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Grace′s Insect Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for insect cell culture