Merck
  • Optoelectronic performance comparison of new thiophene linked benzimidazole conjugates with diverse substitution patterns.
Optoelectronic performance comparison of new thiophene linked benzimidazole conjugates with diverse substitution patterns.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-08-02)
Gözde Murat Saltan, Haluk Dinçalp, Eser Kırmacı, Merve Kıran, Ceylan Zafer
ABSTRACT
In an approach to develop efficient organic optoelectronic devices to be used in light-driven systems, a series of three thiophene linked benzimidazole conjugates were synthesized and characterized. The combination of two thiophene rings to a benzimidazole core decorated with different functional groups (such as OCH
Sigma-Aldrich
3-Methoxyphenylboronic acid
Sigma-Aldrich
2,1,3-Benzothiadiazole, 98%
Sigma-Aldrich
3-(Trifluoromethyl)phenylboronic acid, ≥95%
Sigma-Aldrich
5-Formyl-2-thienylboronic acid, ≥95.0%