Merck
  • Hydrolysis of the amide bond in histidine- and methionine-containing dipeptides promoted by pyrazine and pyridazine palladium(II)-aqua dimers: Comparative study with platinum(II) analogues.
Hydrolysis of the amide bond in histidine- and methionine-containing dipeptides promoted by pyrazine and pyridazine palladium(II)-aqua dimers: Comparative study with platinum(II) analogues.
Bioorganic chemistry (2017-05-02)
Marija D Živković, Snežana Rajković, Biljana Đ Glišić, Nenad S Drašković, Miloš I Djuran
ABSTRACT
Two dinuclear palladium(II) complexes, [{Pd(en)Cl}
Sigma-Aldrich
Pyridazine, 98%
Sigma-Aldrich
Met-Gly