Merck
  • Latency associated peptide has in vitro and in vivo immune effects independent of TGF-beta1.
Latency associated peptide has in vitro and in vivo immune effects independent of TGF-beta1.
PloS one (2008-04-09)
Naeem A Ali, Alice A Gaughan, Charles G Orosz, Chris P Baran, Sara McMaken, Yijie Wang, Timothy D Eubank, Melissa Hunter, Frank J Lichtenberger, Nicholas A Flavahan, Jack Lawler, Clay B Marsh
ABSTRACT
Latency Associated Peptide (LAP) binds TGF-beta1, forming a latent complex. Currently, LAP is presumed to function only as a sequestering agent for active TGF-beta1. Previous work shows that LAP can induce epithelial cell migration, but effects on leukocytes have not been reported. Because of the multiplicity of immunologic processes in which TGF-beta1 plays a role, we hypothesized that LAP could function independently to modulate immune responses. In separate experiments we found that LAP promoted chemotaxis of human monocytes and blocked inflammation in vivo in a murine model of the delayed-type hypersensitivity response (DTHR). These effects did not involve TGF-beta1 activity. Further studies revealed that disruption of specific LAP-thrombospondin-1 (TSP-1) interactions prevented LAP-induced responses. The effect of LAP on DTH inhibition depended on IL-10. These data support a novel role for LAP in regulating monocyte trafficking and immune modulation.
Sigma-Aldrich
Arg-Gly-Glu-Ser acetate salt, ≥95% (HPLC)
Sigma-Aldrich
SB 431542 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Gly-Arg-Gly-Asp-Ser, ≥97% (HPLC)
Sigma-Aldrich
Gey′s Balanced Salt Solution, liquid, sterile-filtered, suitable for cell culture