Merck
  • Occurrence of brominated disinfection byproducts in the air and water of chlorinated seawater swimming pools.
Occurrence of brominated disinfection byproducts in the air and water of chlorinated seawater swimming pools.
International journal of hygiene and environmental health (2017-02-16)
Tarek Manasfi, Brice Temime-Roussel, Bruno Coulomb, Laurent Vassalo, Jean-Luc Boudenne
ABSTRACT
An undesirable consequence of disinfection is the formation of chemical contaminants known as disinfection byproducts (DBPs). Chronic exposure to DBPs has been linked to adverse health effects. The occurrence of DBPs in chlorinated pools filled with seawater (such as thalassotherapy pools and pools in spas) has received little attention so far. The present study evaluated the speciation and levels of disinfection byproducts in indoor swimming pools filled with seawater and treated with chlorine. Water and air samples were collected from three indoor swimming pools located in Southern France. Several classes of DBPs including trihalomethanes, haloacetic acids, haloacetonitriles, and trihaloacetaldehydes were analyzed in water. Halogenated volatile organic compounds were analyzed in air. Extractable organic halides (EOX) contents were determined using combustion/micro-coulometry system. The speciation of DBPs identified in the three pools was predominantly brominated. The mean (arithmetic) concentration of bromoform, dibromoacetic acid, tribromoacetic acid, dibromoacetonitrile and bromal hydrate in the three pools was 79.2, 72.9, 59.9, 26.9 and 10.0μg/L, respectively. By weight, HAAs represented the most abundant chemical class followed by THMs. In air, bromoform was the most abundant THM occurring at a mean concentration of 133.2μg/m
Sigma-Aldrich
2,4,6-Tribromophenol, 99%
Sigma-Aldrich
2,3-Dibromopropionic acid, 98%
Sigma-Aldrich
Tribromoacetaldehyde, 97%